missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-801 Foam Nitrile Coated Nylon Gloves

Catalog No. p-9012026
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19160411A 6
19160411B 7
19160411C 8
19160411D 9
19160411E 10
19160411F 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19160411A Supplier Ansell Supplier No. 1180111060
Only null left

Foam nitrile coated gloves offer an improved design that is up to 30% more breathable to enhance user comfort. The dark colored liner is suitable for slightly dirty or oily environments.

  • Dark colored liner is well suited for slightly dirty or oily environments
  • Foam nitrile coating is up to 30% more breathable for increased user comfort
  • Proprietary washing process efficiently extracts impurities to deliver a cleaner, more comfortable glove
  • Independently certified as skin friendly by the Oeko-Tex™ testing & certification system
  • No transfer of silicone contaminants to metal parts prior to painting

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Dip-coated Gloves
Size 6
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Series 11-801
Yarn Gauge 15
Show More Show Less