Learn More
Ansell HyFlex™ 11-757 Cut-Resistant Touchscreen Gloves
Ultralight ANSI A7 / ISO F-level cut-resistant touchscreen glove.
Supplier: Ansell 11757080VP
Description
- Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility
- Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
- High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
- Tailored fit at the base of the pinky finger better matches natural hand contours
- Dermatologically approved by Dermatest™
Specifications
| Polyurethane | |
| ANSI A7 | |
| Nylon, Stainless Steel Fiber, Polyester, Tungsten |
| Black | |
| Yes | |
| 8 |