Learn More
Ansell HyFlex™ 11-755 Cut-Resistant Touchscreen Gloves
Ultralight ANSI A5/EN ISO E-level cut-resistant gloves with touchscreen compatibility.
$170.86 - $170.86
Specifications
| Coating Material | Polyurethane |
|---|---|
| Color | Gray |
| Cut Resistance Rating | ANSI A5 |
| Material | HPPE, Tungsten, Spandex, Polyester |
| Style | HyFlex™ 11-755 |
Description
- Features INTERCEPT Technology
- Ultra-thin breathable gloves that offer high cut resistance, while maintaining enhanced flexibility and tactility
- Touchscreen compatible
- Designed to fit the natural shape of the hand to help reduce hand fatigue
- Clean and skin friendly. Dermatologically approved by Dermatest
Specifications
| Polyurethane | |
| ANSI A5 | |
| HyFlex™ 11-755 |
| Gray | |
| HPPE, Tungsten, Spandex, Polyester |