Learn More
Ansell HyFlex™ 11-754 Cut-Resistant Industrial Working Gloves, Vend Pack
Ultralight ANSI A4/EN ISO D-rated cut-resistant gloves with touchscreen compatibility.
Supplier: Ansell 11754090VEND
Description
- Ultra-lightweight design provides dexterity for handling and assembly of sharp parts
- Touchscreen compatible glove for enhanced productivity
- Designed to fit the natural shape of the hand to help reduce hand fatigue
- Clean and skin friendly. Dermatologically approved by Dermatest
Specifications
| Polyurethane | |
| ANSI A4 | |
| 9 | |
| Vend Pack |
| Gray | |
| Nylon, Spandex, Stainless Steel Fiber | |
| HyFlex™ 11-754 |