Learn More
Ansell HyFlex™ 11-754 Cut-Resistant Industrial Working Gloves, Vend Pack
Description
- Ultra-lightweight design provides dexterity for handling and assembly of sharp parts
- Touchscreen compatible glove for enhanced productivity
- Designed to fit the natural shape of the hand to help reduce hand fatigue
- Clean and skin friendly. Dermatologically approved by Dermatest
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Gray |
| Cut Resistance Rating | ANSI A4 |
| Material | Nylon, Spandex, Stainless Steel Fiber |
| Size | 7 |
| Style | HyFlex™ 11-754 |
| Packaging Type | Vend Pack |