missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology

Catalog No. 19050082F
Change view
Click to view available options
Size:
10
11
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19050082F 11
19050082C 8
19050082D 9
19050082E 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19050082F Supplier Ansell Supplier No. 11751110

missing translation for 'validMainframeMsg'

Level C cut protection gloves offer enhanced abrasion resistance for extended wear.

  • INTERCEPT Technology™ for high level of cut protection
  • High durability PU coating, increased levels of abrasion resistance
  • Dirt masking design allows for extended product use
  • In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
  • Also available in vendor pack

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HPPE, Fiberglass, Nylon, Spandex, Polyurethane
Product Type Cut-resistant Gloves
Size 11
EN388 Standard EN 388:4X43C
For Use With (Application) General maintenance, hardware assembly, machining, stamping, cutting, raw material handling
Glove Cuff Style Knit
Liner Material HPPE, Fiberglass, Nylon, Spandex
Model 11751110
Quantity 12 Pairs/Bag, 12 Bags/Cs.
Series HyFlex™ 11-751
Length (English) 8.8 in.
Length (Metric) 22.4 cm
Show More Show Less