missing translation for 'onlineSavingsMsg'
Learn More

Hydroxytacrine maleate salt, Thermo Scientific Chemicals

$128.73 - $128.73

Chemical Identifiers

CAS 118909-22-1
Molecular Formula C17H18N2O5
Molecular Weight (g/mol) 330.34
MDL Number MFCD00069287
InChI Key NEEKVKZFYBQFGT-BTJKTKAUSA-N
Synonym velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158
PubChem CID 5702293
IUPAC Name 9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid
SMILES C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60680MD
View Documents
Thermo Scientific Chemicals
J60680MD
250 mg
Each for $128.73
Only null left
 
Description

Description

Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cholinesterase inhibitor
  • Potential Alzheimer's disease therapeutic
Specifications

Chemical Identifiers

118909-22-1
330.34
NEEKVKZFYBQFGT-BTJKTKAUSA-N
5702293
C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O
C17H18N2O5
MFCD00069287
velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158
9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid

Specifications

118909-22-1
White
MFCD00069287
velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158
C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O
330.34
330.34
Hydroxytacrine maleate salt
171°C to 173°C
C17H18N2O5
250 mg
NEEKVKZFYBQFGT-BTJKTKAUSA-N
9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid
5702293
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only