missing translation for 'onlineSavingsMsg'
Learn More

Hydroxytacrine maleate salt, Thermo Scientific Chemicals

Catalog No. AAJ60680MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60680MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60680MD Supplier Thermo Scientific Chemicals Supplier No. J60680MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118909-22-1 | Molecular Formula: C17H18N2O5 | Molecular Weight: 330.34 g/mol

Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cholinesterase inhibitor
  • Potential Alzheimer's disease therapeutic

Chemical Identifiers

CAS 118909-22-1
Molecular Formula C17H18N2O5
Molecular Weight (g/mol) 330.34
MDL Number MFCD00069287
InChI Key NEEKVKZFYBQFGT-BTJKTKAUSA-N
Synonym velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158
PubChem CID 5702293
IUPAC Name 9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid
SMILES C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 171°C to 173°C
Color White
Quantity 250 mg
Formula Weight 330.34
Physical Form Solid
Chemical Name or Material Hydroxytacrine maleate salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only