missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Methylcellulose, 50 cps, FCC, 19-30%, Spectrum™ Chemical

Catalog No. p-7157543
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603352 125 g Poly Bottle
18603353 500 g Poly Bottle
18603354 2.5 kg Poly Pail
18603355 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603352 Supplier Spectrum Chemical Mfg Cor Supplier No. H1072125GM
Only null left

H1072, 9004-65-3

Spectrum™ Chemical Hydroxypropyl Methylcellulose, 50 cps, FCC is most commonly used as an alternative to gelatin and gluten and is approved by FDA as both a direct and an indirect food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
InChI Key PUSNGFYSTWMJSK-UHFFFAOYNA-N
IUPAC Name 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 19 to 30%
Grade FCC
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Poly Bottle