missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Methylcellulose, 4,000 cP, FCC, 19-30%, Spectrum™ Chemical

Catalog No. 18603349
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603349 500 g Poly Pail
18603350 2.5 kg Poly Pail
18603351 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603349 Supplier Spectrum Chemical Mfg Cor Supplier No. H1102500GM

H1102, 9004-65-3

Spectrum™ Chemical Hydroxypropyl Methylcellulose, 4,000 cP, FCC is most commonly used as an alternative to gelatin and gluten and is approved by FDA as both a direct and an indirect food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
InChI Key PUSNGFYSTWMJSK-UHFFFAOYNA-N
IUPAC Name 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 19 to 30%
Grade FCC
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Poly Pail