missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)

CAS: 9004-65-3 | C56H108O30 | 1261.45 g/mol

$220.51 - $220.51

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
MDL Number MFCD00131360
InChI Key PUSNGFYSTWMJSK-GSZQVNRLSA-N
PubChem CID 57503849
IUPAC Name (2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan-
SMILES CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC244021000
SDS View Documents
Thermo Scientific Chemicals
244021000
100 g Plastic bottle
Each for $220.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9004-65-3
1261.45
PUSNGFYSTWMJSK-GSZQVNRLSA-N
(2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan-
C56H108O30
MFCD00131360
57503849
CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

White to Cream
9004-65-3
MFCD00131360
15, 4880
100 g
CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
1261.45
Authentic
Plastic bottle
Hydroxypropyl methyl cellulose
Powder
C56H108O30
1.3900g/mL
Solubility in water: soluble. Other solubilities: insoluble in hot water;, soluble in dmf, dmso, ethylene glycol (hot),, pyridine;, insoluble in anhydrous ethanol, diethyl ether,, chloroform
PUSNGFYSTWMJSK-GSZQVNRLSA-N
(2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan-
57503849
10% max.
1.39
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NF9125000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only