missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hydroxypropyl methyl cellulose, M.N. 86,000, viscosity 4,000 cP (2% solution)

Catalog No. AC244021000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC244021000 100 g Plastic bottle
AC244022500 250 g Plastic bottle
AC244020010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC244021000 Supplier Thermo Scientific Chemicals Supplier No. 244021000

CAS: 9004-65-3 | C56H108O30 | 1261.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-65-3
Molecular Formula C56H108O30
Molecular Weight (g/mol) 1261.45
MDL Number MFCD00131360
InChI Key PUSNGFYSTWMJSK-GSZQVNRLSA-N
PubChem CID 57503849
IUPAC Name (2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan-
SMILES CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White to Cream
Physical Form Powder
Density 1.3900g/mL
Merck Index 15, 4880
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in hot water;, soluble in dmf, dmso, ethylene glycol (hot),, pyridine;, insoluble in anhydrous ethanol, diethyl ether,, chloroform
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Plastic bottle
Specific Gravity 1.39
Chemical Name or Material Hydroxypropyl methyl cellulose
Show More Show Less
RTECSNumber NF9125000
TSCA TSCA

RUO – Research Use Only