missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol

Catalog No. p-7023922
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17951AK 250 mL
AAA17951AU 1000 mL
AAA179510J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17951AK Supplier Thermo Scientific Chemicals Supplier No. A17951AK
Only null left

CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol

Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 27813-02-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004536
InChI Key ZMARGGQEAJXRFP-UHFFFAOYNA-N
Synonym 2-hydroxypropyl methacrylate, 2-hydroxypropylmethacrylate, hpma, beta-hydroxypropyl methacrylate, acryester hp, 2-hydroxypropyl 2-methylacrylate, poly 2-hydroxypropyl methacrylate, 2-hydroxypropyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, 2-hydroxypropyl ester, 2-hpma
PubChem CID 13539
ChEBI CHEBI:53440
IUPAC Name 2-hydroxypropyl 2-methylprop-2-enoate
SMILES CC(CO)OC(=O)C(C)=C

Specifications

Melting Point 70°C (1mmHg)
Density 1.029
Boiling Point 205°C to 209°C
Flash Point 97°C (207°F)
Refractive Index 1.448
Quantity 250 mL
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Hydroxypropyl methacrylate, mixture of isomers, stabilized with ≈0.02% 4-methoxyphenol
Show More Show Less
Hazard Category H317-H319
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
EINECSNumber 248-666-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only