missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl cellulose, M.W. 100,000

Catalog No. p-7058872
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4340014 25 g
AA4340022 100 g
AA4340036 500 g
AA43400A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4340014 Supplier Thermo Scientific Chemicals Supplier No. 04340014
Only null left

Thickener, binder, and emulsifier | CAS: 9004-64-2 | (C24H44O16)n

Thickener, binder, and emulsifier.Hydroxypropyl cellulose is used as an emulsifier, stabilizer, whipping aid and thickener. It is also used as a binder in ceramics and glazes, additive in hair and cosmetic products and blow-molded bottles. Further, it is used in contact lens wetting solutions, as a formulation aid and texturizer for foods. In addition, it serves as a topical ophthalmic protectant and lubricant. It acts as a disintegrant and a binder in tablets. It is also utilized as a sieving matrix for DNA separations by capillary and microchip electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c

Specifications

pH 7
Odor Odorless
Quantity 25 g
Merck Index 14,4841
Solubility Information Soluble in polar organic solvents,in water at room temperature;Soluble in water,methyl alcohol,ethyl alcohol and other organic solvents.
Physical Form Powder
Chemical Name or Material Hydroxypropyl cellulose, M.W. 100,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only