missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000

Catalog No. p-147545
Change view
Click to view available options
Quantity:
100 g
250 g
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184881000 100 g Glass bottle
AC184882500 250 g Glass bottle
AC184880051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC184881000 Supplier Thermo Scientific Chemicals Supplier No. 184881000
Only null left

CAS: 9004-64-2 | (C24H44O16)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c

Specifications

Color White to Cream
pH 5 to 8.5 (1% soln.)
Physical Form Granules or Crystalline Powder
Quantity 100 g
Formula Weight 100.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
Packaging Glass bottle
Viscosity 75-150 mPa.s 5% aq.sol.
Chemical Name or Material Hydroxypropyl cellulose
RTECSNumber NF9050000
TSCA TSCA

RUO – Research Use Only