missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hydroxypropyl cellulose, Average M.W. 100.000
Supplier: Thermo Scientific™ 184880010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Hydroxypropyl cellulose | |
| (C24H44O16)n | |
| 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c | |
| 100.000 (Average ) | |
| 5% max. (105°C) | |
| 75-150 mPa.s 5% aq.sol. |
| 9004-64-2 | |
| MFCD00132688 | |
| 1kg | |
| Authentic | |
| Glass bottle |
Safety and Handling
RTECSNumber : NF9050000
TSCA : TSCA
RUO â Research Use Only