missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose, 75-150 cP, NF, 53.4-850%, Spectrum™ Chemical

Catalog No. 18603340
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603340 125 g Poly Bottle
18603341 500 g Poly Pail
18603342 2.5 kg Poly Pail
18603343 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603340 Supplier Spectrum Chemical Mfg Cor Supplier No. HY117125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

HY117, 9004-64-2

Spectrum™ Chemical Hydroxypropyl Cellulose, 75-150 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
SMILES CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*

Specifications

CAS Max % 100%
Ignition Residue 0.8%
Loss on Drying 5%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 53.4 to 80.5%
Grade NF