missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxypropyl Cellulose (3-6mPa.s, 2% in Water at 20°C), TCI America™
Supplier: TCI America H047325G
Specifications
| Hydroxypropyl Cellulose (3-6mPa.s, 2% in Water at 20°C) | |
| White-Yellow | |
| (C24H44O16)n | |
| 25 g | |
| Crystalline Powder |
| 9004-64-2 | |
| 5.0 to 7.5 (2% H2O soln.) | |
| MFCD00132688 | |
| 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c |
Chemical Identifiers
| 9004-64-2 | |
| (C24H44O16)n | |
| MFCD00132688 | |
| 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c |
Safety and Handling
EINECSNumber : (8)-0194
RTECSNumber : NF9050000
TSCA : Yes