missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose (1,000-4,000mPa.s, 2% in Water at 20°C), TCI America™

Catalog No. H047525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H047525G 25 g
H0475500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H047525G Supplier TCI America Supplier No. H047525G

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Hydroxypropyl Cellulose (1,000-4,000mPa.s, 2% in Water at 20°C)
EINECSNumber (8)-0194
RTECSNumber NF9050000
TSCA Yes