missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Acrylate (mixture of 2-Hydroxypropyl and 2-Hydroxy-1-methylethyl Acrylate) 90.0+%, TCI America™

Catalog No. a074425ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A074425ML 25 mL
A0744500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A074425ML Supplier TCI America Supplier No. A074425ML
Only null left

(stabilized with MEHQ)

Chemical Identifiers

CAS 25584-83-2
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD04113589
InChI Key QZPSOSOOLFHYRR-UHFFFAOYSA-N
Synonym Acrylic Acid Hydroxypropyl Ester, Propylene Glycol Monoacrylate
PubChem CID 33101
IUPAC Name 3-hydroxypropyl prop-2-enoate
SMILES C=CC(=O)OCCCO

Specifications

Color Yellow
Boiling Point 77°C
Quantity 25 mL
UN Number 2922
Formula Weight 130.14
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Hydroxypropyl Acrylate (mixture of 2-Hydroxypropyl and 2-Hydroxy-1-methylethyl Acrylate) (stabilized
EINECSNumber (2)-0958&(2)-0997
RTECSNumber UD3610000
TSCA Yes