missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxynaphthol Blue, Indicator for Metal Titration, Honeywell Fluka™
Indicator for metal titration
$258.94 - $990.96
Chemical Identifiers
| CAS | 63451-35-4 |
|---|---|
| Molecular Formula | C20H11N2Na3O11S3 |
| Molecular Weight (g/mol) | 620.46 |
| MDL Number | MFCD00036393 |
| InChI Key | JQECMNGWLIFQLL-UHFFFAOYSA-K |
| PubChem CID | 44134769 |
| IUPAC Name | trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate |
| SMILES | C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 63451-35-4 | |
| 620.46 | |
| JQECMNGWLIFQLL-UHFFFAOYSA-K | |
| trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate |
| C20H11N2Na3O11S3 | |
| MFCD00036393 | |
| 44134769 | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+] |
Specifications
| 63451-35-4 | |
| MFCD00036393 | |
| JQECMNGWLIFQLL-UHFFFAOYSA-K | |
| Conforms to structure | |
| 620.46 | |
| 620.47g/mol | |
| Hydroxynaphthol blue |
| C20H11N2Na3O11S3 | |
| NONH for all modes of transport | |
| C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+] | |
| trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate | |
| 44134769 | |
| Liquid |
Safety and Handling
RTECSNumber : 264-197-7