Learn More
Hydroxylapatite, fast flow
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids | CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol
Supplier: Thermo Scientific Chemicals J6007618
Description
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydroxylapatite | |
| 1306-06-5 | |
| Ca5HO13P3 | |
| MFCD00010904 | |
| 14,3467 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate | |
| 502.32 |
| fast flow | |
| White | |
| Ca5(OH)(PO4)3 | |
| 50 g | |
| Calcium phosphate hydroxide; Durapatite | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
| 502.31 | |
| Powder |
Chemical Identifiers
| 1306-06-5 | |
| 502.31 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate |
| Ca5HO13P3 | |
| MFCD00010904 | |
| Calcium phosphate hydroxide; Durapatite | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 215-145-7
RTECSNumber : MY8434000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only