missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylapatite, fast flow

Catalog No. AAJ6007609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6007609 10 g
AAJ6007618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6007609 Supplier Thermo Scientific Chemicals Supplier No. J6007609
Only null left

Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids | CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol

Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide; Durapatite
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Linear Formula Ca5(OH)(PO4)3
Quantity 10 g
Merck Index 14,3467
Formula Weight 502.32
Physical Form Powder
Chemical Name or Material Hydroxylapatite, fast flow
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 215-145-7
RTECSNumber MY8434000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only