missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylapatite, for analysis
CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol
$39.95 - $310.65
Chemical Identifiers
| CAS | 1306-06-5 |
|---|---|
| Molecular Formula | Ca5HO13P3 |
| Molecular Weight (g/mol) | 502.31 |
| MDL Number | MFCD00010904 |
| InChI Key | XYJRXVWERLGGKC-UHFFFAOYSA-D |
| Synonym | Calcium phosphate hydroxide |
| IUPAC Name | pentacalcium hydroxide triphosphate |
| SMILES | [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC371260250
|
Thermo Scientific Chemicals
371260250 |
25 g | Glass bottle |
Each for $39.95
|
|
||||
|
AC371261000
|
Thermo Scientific Chemicals
371261000 |
100 g | Glass bottle |
Each for $136.44
|
|
||||
|
AC371260010
|
Thermo Scientific Chemicals
371260010 |
1 kg | Plastic Bottle |
Each for $310.65
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1306-06-5 | |
| 502.31 | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate |
| Ca5HO13P3 | |
| MFCD00010904 | |
| Calcium phosphate hydroxide | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Specifications
| 1700.0°C | |
| 1306-06-5 | |
| 25 g | |
| 3Ca3(PO4)2·Ca(OH)2 | |
| 15, 3514 | |
| Solubility in water: negligible | |
| [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
| 502.31 | |
| 35 to 40% (Ca) | |
| 8% max. (800°C) | |
| Hydroxylapatite |
| White | |
| Powder | |
| Ca5HO13P3 | |
| MFCD00010904 | |
| Calcium phosphate hydroxide | |
| XYJRXVWERLGGKC-UHFFFAOYSA-D | |
| pentacalcium hydroxide triphosphate | |
| 502.31 | |
| Analytical | |
| Glass bottle |
Safety and Handling
EINECSNumber : 215-145-7
RUO – Research Use Only