missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate 98.0+%, TCI America™

Catalog No. p-7130397
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
H019625G 25 g
H0196500G 500 g
2 options

Catalog No. H019625G

Supplier: TCI America H019625G

Only null left

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H3NO
MDL Number MFCD00044869
Synonym Hydroxylammonium Sulfate

Specifications

Melting Point 170°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 2865
Formula Weight 164.13
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydroxylamine Sulfate
RTECSNumber NC5425000
TSCA Yes