missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine Sulfate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 170°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| UN Number | 2865 |
| Molecular Weight (g/mol) | 164.13 |
| Formula Weight | 164.13 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | Hydroxylamine Sulfate |
Safety and Handling
| RTECSNumber | NC5425000 |
| TSCA | Yes |