missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate 98.0+%, TCI America™

Catalog No. H019625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H019625G 25 g
H0196500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H019625G Supplier TCI America Supplier No. H019625G

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H3NO
MDL Number MFCD00044869
Synonym Hydroxylammonium Sulfate

Specifications

Melting Point 170°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 2865
Formula Weight 164.13
Percent Purity ≥98.0% (T)
Chemical Name or Material Hydroxylamine Sulfate
RTECSNumber NC5425000
TSCA Yes