missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165684
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611001 25 g Poly Bottle
18610996 250 g Poly Bottle
18610999 500 g Poly Bottle
18611000 1 kg Poly Bottle
18610997 2.5 kg Poly Pail
18610995 12 kg Poly Pail
18610998 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18611001 Supplier Spectrum Chemical Mfg Cor Supplier No. H235725GM
Only null left

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H8N2O6S
Molecular Weight (g/mol) 164.13
InChI Key VGYYSIDKAKXZEE-UHFFFAOYSA-L
IUPAC Name bis(hydroxyazanium) sulfate
SMILES [NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Packaging Poly Bottle
Quantity 25 g
Percent Purity 0.99
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.