missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate, 99% Reagent, Reagents

Catalog No. 01337216
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337216 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337216 Supplier Labchem Inc Supplier No. C2216550500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydroxylamine Sulfate, 99% Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H8N2O6S
Molecular Weight (g/mol) 164.13
InChI Key VGYYSIDKAKXZEE-UHFFFAOYSA-L
Synonym Oxammonium Sulfate
IUPAC Name bis(hydroxyazanium) sulfate
SMILES [NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O

Specifications

Melting Point 170°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.88
Chemical Name or Material Hydroxylamine Sulfate, 99% Reagent