missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride 97.0+%, TCI America™

Catalog No. H1581500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1581500G 500 g
H158125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1581500G Supplier TCI America Supplier No. H1581500G

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Melting Point 151°C
Color White
pH 2.5 to 3.5 (50g/L H2O 25°C)
Physical Form Crystalline Powder
Quantity 500 g
UN Number 2923
Formula Weight 69.49
Percent Purity ≥97.0% (T)
Chemical Name or Material Hydroxylamine Hydrochloride
RTECSNumber NC3675000
TSCA Yes