missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride (Suitable for Mercury Determination/Certified ACS), Fisher Chemical™

Catalog No. p-145066
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
H330100 100 g Poly Bottle
H330500 500 g Poly Bottle
H3301 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H330100 Supplier Thermo Fisher Scientific Supplier No. H330 100
Only null left

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Melting Point 155°C
Color White
pH 2.5 to 3.5
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥96 %
Linear Formula NH2OH·HCl
UN Number 2923
Formula Weight 69.49
Percent Purity ≥96%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure Negligible
Density 1.6700g/cm³
Chemical Name or Material Hydroxylamine Hydrochloride, Suitable for Mercury Determination
Show More Show Less
DOTInformation DOT Class 8, : Corrosive