missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, Hygroscopic, ACS Grade, ≥ 96.0% , LabChem™

Catalog No. LC155151
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC155151 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC155151 Supplier LabChem Supplier No. LC155151
Only null left

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Melting Point 152°C
Color Colorless
CAS Max % 100
Physical Form Solid
Quantity 500 g
Linear Formula H3NO·HCl
UN Number UN2923
Solubility Information Soluble in water
Formula Weight 69.49
Percent Purity ≥96.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Density 1,670kg/m3
Chemical Name or Material Hydroxylamine Hydrochloride, Hygroscopic
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if swallowed or in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing mustd not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 226-798-2
Recommended Storage Room Temperature