missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine hydrochloride, ACS, 96+%

Catalog No. AA3641622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3641622 100 g
AA3641636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3641622 Supplier Thermo Scientific Chemicals Supplier No. 03641622

A Monoamine Oxidase Inhibitor; Inhibits Platelet Aggregation | CAS: 5470-11-1 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 152°C (decomposition)
Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range >96%
Beilstein 3539763
Solubility Information Soluble in water,alcohol,methanol,glycerol
Formula Weight 69.49
Grade ACS
Odor Acidic
Sensitivity Hygroscopic
Appearance White solid
Density 1.67
Chemical Name or Material Hydroxylamine hydrochloride, For analysis ACS
Show More Show Less
Hazard Category H290-H301-H312-H315-H317-H319-H351-H373-H500
Precautionary Statement P201-P202-P234-P260-P264b-P270-P272-P280-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 226-798-2
RTECSNumber NC3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only