Learn More
Hydroxylamine hydrochloride, 99+%
CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Supplier: Thermo Scientific Chemicals 270100050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Hydroxylamine hydrochloride | |
| 5470-11-1 | |
| Crystalline Powder, Crystals and/or Chunks | |
| 99+% | |
| H2NOH·HCl | |
| 01,478; 02,217; 07,176; 09,245 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| hydroxylamine;hydrochloride | |
| 443297 | |
| 69.49 | |
| Authentic |
| 155°C to 158°C | |
| White | |
| 5 kg | |
| ClH4NO | |
| MFCD00051089 | |
| 15, 4867 | |
| Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether | |
| Cl.NO | |
| 69.49 | |
| CHEBI:5807 | |
| ≥99% | |
| Plastic drum |
Chemical Identifiers
| 5470-11-1 | |
| 69.49 | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| 443297 | |
| Cl.NO |
| ClH4NO | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| CHEBI:5807 |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing canc
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protecti
GHS Signal Word: Danger
EINECSNumber : 226-798-2
RUO – Research Use Only