missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 50 g + 100 mL Water, Ricca Chemical

Supplier:  Ricca Chemical Company R39250004A

Quantity 4 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. R39250004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

5470-11-1
69.49
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807
NO.Cl
ClH4NO
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
hydroxylamine;hydrochloride

Specifications

Hydroxylamine Hydrochloride
5470-11-1
ClH4NO
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
Natural Poly Bottle
50g + 100mL Water
4 L
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
NO.Cl
69.49
CHEBI:5807
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents