missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyethyl Cellulose (800-1,500mPa.s, 2% in Water at 20°C), TCI America™
$55.07 - $149.98
Chemical Identifiers
| CAS | 9004-62-0 |
|---|---|
| Molecular Formula | C29H52O21 |
| Molecular Weight (g/mol) | 736.714 |
| MDL Number | MFCD00072770 |
| InChI Key | CWSZBVAUYPTXTG-UHFFFAOYSA-N |
| PubChem CID | 24846132 |
| IUPAC Name | 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| SMILES | CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O |
Chemical Identifiers
| 9004-62-0 | |
| 736.714 | |
| CWSZBVAUYPTXTG-UHFFFAOYSA-N | |
| 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| C29H52O21 | |
| MFCD00072770 | |
| 24846132 | |
| CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O |
Specifications
| 9004-62-0 | |
| MFCD00072770 | |
| CC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)COC3C(C(C(C(O3)CO)OC)O)O)OC4C(C(C(C(O4)CO)OC)O)OCCO)O)O)O)O | |
| 736.714 | |
| Crystalline Powder |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYSA-N | |
| 5-[6-[[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)-2-methyloxane-3,4-diol | |
| 24846132 | |
| Hydroxyethyl Cellulose (800-1,500mPa.s, 2% in Water at 20°C) |
Safety and Handling
EINECSNumber : (8)-0194
RTECSNumber : FJ5958000
TSCA : Yes