missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyethyl Cellulose, 5,000 cP, NF, Spectrum™ Chemical
H1236, 9004-62-0
Supplier: Spectrum Chemical Mfg Cor H123612KGBL
| Quantity | 12 kg |
|---|---|
| Packaging | Poly Pail |
Description
Spectrum™ Chemical Hydroxyethyl Cellulose, 5,000 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-62-0 | |
| 736.71 | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO |
Specifications
| 9004-62-0 | |
| 5% | |
| Poly Pail | |
| 12 kg | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO | |
| 736.71 |
| 100% | |
| 10% | |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol | |
| NF |