missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyethyl Cellulose, 5,000 cP, NF, Spectrum™ Chemical

H1236, 9004-62-0

Supplier:  Spectrum Chemical Mfg Cor H1236500GM

Quantity 500 g
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18603314


Only null left

Description

Description

Spectrum™ Chemical Hydroxyethyl Cellulose, 5,000 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

9004-62-0
736.71
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
C29H52O21
CWSZBVAUYPTXTG-UHFFFAOYNA-N
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO

Specifications

9004-62-0
5%
Poly Bottle
500 g
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
736.71
100%
10%
C29H52O21
CWSZBVAUYPTXTG-UHFFFAOYNA-N
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
NF
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions