missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyethyl Cellulose, 3,400 cps, Spectrum™ Chemical
H1148, 9004-62-0
Supplier: Spectrum Chemical Mfg Cor H114812KG
| Quantity | 12 kg |
|---|---|
| Packaging | Poly Pail |
Description
Spectrum™ Chemical Hydroxyethyl Cellulose, 3,400 cps is a thickening and gelling agent created from cellulose, most commonly used in cleaning solutions, cosmetics and other products used in the household. It usually comes in capsule form. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 9004-62-0 | |
| 736.71 | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO |
Specifications
| 9004-62-0 | |
| 5% | |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol | |
| Reagent |
| 100% | |
| Poly Pail | |
| 12 kg | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO | |
| 736.71 |