missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyethyl Cellulose, 3,400 cps, Spectrum™ Chemical

H1148, 9004-62-0

Supplier:  Spectrum Chemical Mfg Cor H11482.5KG

Quantity 2.5 kg
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18603305


Only null left

Description

Description

Spectrum™ Chemical Hydroxyethyl Cellulose, 3,400 cps is a thickening and gelling agent created from cellulose, most commonly used in cleaning solutions, cosmetics and other products used in the household. It usually comes in capsule form. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Chemical Identifiers

9004-62-0
736.71
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
C29H52O21
CWSZBVAUYPTXTG-UHFFFAOYNA-N
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO

Specifications

9004-62-0
5%
C29H52O21
CWSZBVAUYPTXTG-UHFFFAOYNA-N
5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol
Reagent
100%
Poly Bottle
2.5 kg
COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO
736.71
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions