missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyethyl Cellulose, 2,000 cP, NF, Spectrum™ Chemical
H2694, 9004-62-0
$898.03 - $10,550.88
Chemical Identifiers
| CAS | 9004-62-0 |
|---|---|
| Molecular Formula | C29H52O21 |
| Molecular Weight (g/mol) | 736.71 |
| InChI Key | CWSZBVAUYPTXTG-UHFFFAOYNA-N |
| IUPAC Name | 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| SMILES | COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603299
|
N/A | 500 g | Poly Bottle |
Each for $898.03
|
|
||||
|
18603300
|
N/A | 2.5 kg | Poly Pail |
Each for $2,516.61
|
|
||||
|
18603301
|
N/A | 12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Hydroxyethyl Cellulose, 2,000 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-62-0 | |
| 736.71 | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO |
Specifications
| 9004-62-0 | |
| 5% | |
| C29H52O21 | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO | |
| 736.71 |
| 100% | |
| 10% | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol | |
| NF |