missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyethyl Cellulose, 100 cP, NF, Spectrum™ Chemical
HY112, 9004-62-0
Supplier: Spectrum Chemical Mfg Cor HY11212KG
| Quantity | 12 kg |
|---|---|
| Packaging | Poly Pail |
Description
Spectrum™ Chemical Hydroxyethyl Cellulose, 100 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-62-0 | |
| 736.71 | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO |
Specifications
| 9004-62-0 | |
| 5% | |
| Poly Pail | |
| 12 kg | |
| COC1C(O)C(O)C(OCC2OC(OC3C(O)C(O)C(C)OC3CO)C(O)C(O)C2OC2OC(CO)C(OC)C(O)C2OCCO)OC1CO | |
| 736.71 |
| 100% | |
| 10% | |
| C29H52O21 | |
| CWSZBVAUYPTXTG-UHFFFAOYNA-N | |
| 5-{[6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}methyl)-3,4-dihydroxy-5-{[4-hydroxy-3-(2-hydroxyethoxy)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy}oxan-2-yl]oxy}-6-(hydroxymethyl)-2-methyloxane-3,4-diol | |
| NF |