missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Sulfate 98.0+%, TCI America™

Catalog No. p-7130969
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13065G 5 g
H130625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13065G Supplier TCI America Supplier No. H13065G
Only null left

Chemical Identifiers

CAS 747-36-4
Molecular Formula C18H28ClN3O5S
Molecular Weight (g/mol) 433.948
MDL Number MFCD00078203
InChI Key JCBIVZZPXRZKTI-UHFFFAOYSA-N
Synonym hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788
PubChem CID 12947
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
SMILES CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 433.95
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxychloroquine Sulfate
RTECSNumber KK3342500
TSCA No