missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Sulfate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022857
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022857 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022857 Supplier Merck Emd Millipore Supplier No. BP1100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydroxychloroquine Sulfate is a salt of hydroxychloroquine (HCQ, Plaquenil), a 4-aminoquinoline based antiviral drug. Product provided as delivered and specified by the issuing Pharmacopoeia. All supporting information has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 747-36-4
Melting Point 238°C to 241°C
Molecular Formula C18H26ClN3O · H2SO4
Linear Formula C18H26ClN3O · H2SO4
MDL Number MFCD00078203
Quantity 100 mg
Synonym 7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate
Formula Weight 433.95
Grade BP
Physical Form Solid
Show More Show Less