missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Naphthol Blue, Solid Dilution (1:133 with Sodium Chloride), Reagent, ACS, Spectrum™ Chemical

Catalog No. 18610972
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18610972 5 g
18610973 25 g
18610974 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610972 Supplier Spectrum Chemical Mfg Cor Supplier No. H11375GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Specifications

Chemical Name or Material Hydroxy Naphthol Blue
CAS 63451-35-4, 7647-14-5, 165660-27-5
Concentration 1:133
Grade Chemical Solution
Packaging Amber Glass Bottle
Color Purple
Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.