missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Hydroxy-2-naphthylmethyl)phosphonic acid, 98%, Thermo Scientific Chemicals
Description
A selective insulin receptor tyrosine kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 5 mg |
| Formula Weight | 238.18 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | (Hydroxy-2-naphthylmethyl)phosphonic acid |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only