missing translation for 'onlineSavingsMsg'
Learn More

Hydroxocobalamin, USP, 95-102%, Spectrum™ Chemical

Catalog No. p-7157553
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603285 5 g Amber Glass Bottle
18603286 25 g Amber Glass Bottle
18603287 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603285 Supplier Spectrum Chemical Mfg Cor Supplier No. H11295GM
Only null left

H1129, 13422-51-0, C62H89CoN13O15P

Spectrum™ Chemical Hydroxocobalamin, USP is a natural form of vitamin B12 and used to treat vitamin deficiency and for treatment for cyanide poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13422-51-0
Molecular Formula C62H90CoN13O15P
Molecular Weight (g/mol) 1347.39
InChI Key PJOHJFIDLYDPIZ-PMEYKKDOSA-M
IUPAC Name λ²-cobalt(2+) (2R,3S,4S,6Z,8S,9S,11Z,14S,16Z,18R,19R)-4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-(2-{[(2S)-2-{[(2R,3S,4R,5S)-5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl]carbamoyl}ethyl)-2,3,6,8,13,13,16,18-octamethyl-20λ⁵,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaene-20,20-diid-20-yl λ³-oxidanide
SMILES [OH2-].[Co++].C[C@@H](CNC(=O)CC[C@]1(C)[C@@H](CC(N)=O)C2[N--]\C1=C(C)/C1=N/C(=C\C3=N\C(=C(C)/C4=N[C@]2(C)[C@@](C)(CC(N)=O)[C@@H]4CCC(N)=O)\[C@@](C)(CC(N)=O)[C@@H]3CCC(N)=O)/C(C)(C)[C@@H]1CCC(N)=O)OP([O-])(=O)O[C@@H]1[C@@H](CO)O[C@@H]([C@@H]1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 95 to 102%
Grade USP