missing translation for 'onlineSavingsMsg'
Learn More

Hydroxocobalamin Acetate 90.0+%, TCI America™

Catalog No. H15421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15421G 1 g
H15425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15421G Supplier TCI America Supplier No. H15421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22465-48-1
Molecular Formula C64H92CoN13O16P
Molecular Weight (g/mol) 1389.42
MDL Number MFCD00082532
InChI Key YKDAZHZZVTXKEY-FIJLCNGJSA-M
Synonym hydroxocobalamin acetate
PubChem CID 91972123
IUPAC Name λ²-cobalt(2+) (2R,3S,4S,6Z,8S,9S,11Z,16Z,18R)-4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[(2R,3S,4R,5S)-5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20λ⁵,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaene-20,20-diid-20-yl hydrogen ethanehydrogen carboxylate
SMILES [Co++].CC([OH-])=O.CC(CNC(=O)CC[C@]1(C)C(CC(N)=O)C2[N--]\C1=C(C)/C1=N/C(=C\C3=N\C(=C(C)/C4=N[C@]2(C)[C@@](C)(CC(N)=O)[C@@H]4CCC(N)=O)\[C@@](C)(CC(N)=O)[C@@H]3CCC(N)=O)/C(C)(C)C1CCC(N)=O)OP([O-])(=O)O[C@@H]1[C@@H](CO)O[C@@H]([C@@H]1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

Quantity 1 g
Formula Weight 1388.41
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxocobalamin Acetate
RTECSNumber GG3740000
TSCA No
Recommended Storage Refrigerator