missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Hydrol Pre-filled Containers, 10% NBF
Hydrol containers are made of polypropylene and are bulk packaged in corrugate trays. These are packaged in larger quantities and are ideal for larger labs with high throughput and have the ability to store large amounts of containers.
Supplier: Epredia™ LC0060
Specifications
| Pre-filled Specimen Container | |
| 60 mL | |
| CH2O | |
| 300/Cs. | |
| C=O | |
| 30.03 |
| Polypropylene | |
| 10% Formalin | |
| MFCD00003274 | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| formaldehyde |