missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Hydrol Pre-filled Containers, 10% NBF

Hydrol containers are made of polypropylene and are bulk packaged in corrugate trays. These are packaged in larger quantities and are ideal for larger labs with high throughput and have the ability to store large amounts of containers.

Supplier:  Epredia™ RBBP0240

 View more versions of this product

Catalog No. 22045405


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pre-filled Specimen Container
240 mL
CH2O
50/Cs.
C=O
30.03
Polypropylene
10% Formalin
MFCD00003274
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions